missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TGF beta-3 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543704
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TGF beta-3 Monoclonal Antibody for Western Blot
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
Specifikationer
| TGF beta-3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10600 | |
| TGFB3 | |
| Recombinant protein Transforming Growth Factor Beta 3. The antigen corresponds to amino acid range 24-300 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| TGFB3 | |
| ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering