missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TGFBR3 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899484
|
Proteintech
20000-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16889484
|
Proteintech
20000-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to TGFBR3.
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
TGFBR3 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q03167 | |
7049 | |
Peptide | |
Primary | |
-20°C | |
TGFBR3 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
betaglycan, BGCAN, TGF beta receptor type 3, TGF beta receptor type III, TGF-beta Receptor III, TGFBR3, TGFR 3, TGFbeta RIII | |
TGFBR3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |