Läs mer
TH-237A, MedChemExpress
Produktkod. 16457107
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16457107 5 mg 5mg
16435286 100 mg -
16430707 50 mg -
16414388 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16457107 Leverantör Medchem Express Leverantörsnummer HY11054
MedChemExpress TH-237A(meso-GS 164) is a novel neuroprotective agent exhibiting favorable permeation across the blood brain barrier.
TH-237A(meso-GS 164) is a novel neuroprotective agent exhibiting favorable permeation across the blood brain barrier.
Specifikationer
Kemiskt namn eller material TH-237A
CAS 935467-97-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C18H17F2NO3
Kvantitet 5 mg
Synonym meso-GS 164
Löslighetsinformation Ethanol : 100 mg/mL (300.00 mM; Need ultrasonic) ∣DMSO : ≥ 46 mg/mL (138.00 mM)
LEDER OCC12N([C@H](C3=CC=C(F)C=C3)OC2)[C@@H](C4=CC=C(F)C=C4)OC1
Molekylvikt (g/mol) 333.33
Formel vikt 333.33
Procent renhet 98.55%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration