missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ THAL SNS 032
GREENER_CHOICE

Produktkod. 18729286
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18729286 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18729286 Leverantör Tocris Bioscience™ Leverantörsnummer 6532/5

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Potent and selective Cdk9 Degrader (PROTAC™)

PROTAC comprising the cyclin-dependent kinase inhibitor SNS 032 (Cat. No. 4075) conjugated to the cereblon E3 ligase ligand, thalidomide (Cat. No. 0652). Potent, selective and cereblon-dependent degrader of Cdk9 (EC50 = 4 nM). Displays >15-fold selectivity for Cdk9 over other CDKs (EC50 values are 62, 171 and 398 nM for Cdk2, Cdk1 and Cdk7, respectively). Induces complete degradation of Cdlk9 at 250 nM in MOLT4 cells. Inhibits proliferation of leukemia cell lines.

THAL SNS 032 is a degrader (PROTAC®) comprising the cyclin-dependent kinase inhibitor SNS 032 (Cat. No. 4075) conjugated to the cereblon E3 ligase ligand, thalidomide (Cat. No. 0652). Potent, selective and cereblon-dependent degrader of Cdk9 (EC50 = 4 nM). Displays >15-fold selectivity for Cdk9 over other CDKs (EC50 values are 62, 171 and 398 nM for Cdk2, Cdk1 and Cdk7, respectively). Induces complete degradation of Cdk9 at 250 nM in MOLT4 cells. Inhibits proliferation of leukemia cell lines.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)-1-(14-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-2-oxo-6,9,12-trioxa-3-azatetradecyl)piperidine-4-carboxamide
CAS 2139287-33-3
Kvantitet 5 mg
Molekylformel C40H52N8O10S2
Formel vikt 869.02
Renhet 98%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.