Läs mer
Tocris Bioscience™ Thalidomide 4'-oxyacetamide-PEG3-amine
Beskrivning
Thalidomide 4'-oxyacetamide-PEG3-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 1
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| CAS | 2245697-84-9 |
| Synonym | Thalidomide - linker 1 |
| Kvantitet | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylformel | C23H30N4O9.HCl |
| Formel vikt | 542.97 |
| Renhet | 95% |