missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ Thalidomide, 5'-fluoro

Beskrivning
Thalidomide 5'- fluoro is a derivative of Thalidomide (Cat. No. 0652), commonly used as a precursor to PROTAC® Degraders that hijacks cereblon as the E3 ubiquitin ligase component. Supplied with a fluoro functional handle at a position known not to significantly affect binding to cereblon, ready for conjugation to a linker/target protein ligand.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Require a different derivative? Get in touch .
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-(2,6-Dioxo-3-piperidinyl)-5-fluoro-1 H-isoindole-1,3(2 H)-dione |
| CAS | 835616-61-0 |
| Kvantitet | 100 mg |
| Mål | Degrader Building Blocks |
| Molekylformel | C13H9FN2O4 |
| Renhet | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?