missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ Thalidomide, propargyl
GREENER_CHOICE

Produktkod. 18797256
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18797256 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18797256 Leverantör Tocris Bioscience™ Leverantörsnummer 6685/50

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Alkyne-functionalized Thalidomide; 'click' cereblon ligand for PROTAC™s

Thalidomide, propargyl is a click-activated Thalidomide (Cat.No. 0652); can be used as a precursor to a PROTAC® that hijacks cereblon as the E3 ubiquitin ligase component. Supplied with an alkyne functional handle at a position known not to significantly affect binding to cereblon, for ready click conjugation to a linker/target protein ligand.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Require a different derivative? Get in touch .

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2-(2,6-Dioxo-3-piperidinyl)-4-(2-propyn-1-yloxy)-1H-isoindole-1,3(2H)-dione
CAS 2098487-39-7
Kvantitet 50 mg
Molekylformel C16H12N2O5
Formel vikt 312.28
Produkttyp Alkyne-functionalized Ligand
Renhet 95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.