Läs mer
Theaflavin, TRC
Produktkod. 30453919
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453919 1 mg
30449542 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30453919 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3402001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4670-05-7
Molekylformel C29 H24 O12
Molekylvikt (g/mol) 564.49
Synonym 5H-Benzocyclohepten-5-one, 1,8-bis[(2R,3R)-3,4-dihydro-3,5,7-trihydroxy-2H-1-benzopyran-2-yl]-3,4,6-trihydroxy-, 5H-Benzocyclohepten-5-one, 1,8-bis(3,4-dihydro-3,5,7-trihydroxy-2H-1-benzopyran-2-yl)-3,4,6-trihydroxy-, [2R-[2α(2R*,3R*),3α]]-, 5H-Benzocyclohepten-5-one, 3,4,6-trihydroxy-1,8-bis(3α,5,7-trihydroxy-2α-chromanyl)- (8CI), Theaflavin (7CI), Theaflavine (6CI), (-)-Theaflavin, 1,8-Bis[(2R,3R)-3,4-dihydro-3,5,7-trihydroxy-2H-1-benzopyran-2-yl]-3,4,6-trihydroxy-5H-benzocyclohepten-5-one, Theaflavin 1, Xiayie
IUPAC-namn 3,4,6-trihydroxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-2-yl]benzo[7]annulen-5-one
LEDER O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1C3=Cc4c(cc(O)c(O)c4C(=O)C(=C3)O)[C@H]5Oc6cc(O)cc(O)c6C[C@H]5O
Specifikationer
Kemiskt namn eller material Theaflavine
Kvantitet 1 mg
InChI formel InChI=1S/C29H24O12/c30-11-3-17(32)15-8-21(36)28(40-23(15)5-11)10-1-13-14(7-20(35)27(39)25(13)26(38)19(34)2-10)29-22(37)9-16-18(33)4-12(31)6-24(16)41-29/h1-7,21-22,28-33,35-37,39H,8-9H2,(H,34,38)/t21-,22-,28-,29-/m1/s1
Formel vikt 564.1268