Läs mer
Thenium Closylate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thenium Closylate |
| CAS | 4304-40-9 |
| Molekylformel | C15 H20 N O S . C6 H4 Cl O3 S |
| Kvantitet | 250 mg |
| Synonym | 2-Thiophenemethanaminium','N,N-dimethyl-N-(2-phenoxyethyl)-','4-chlorobenzenesulfonate (1:1), 2-Thiophenemethanaminium','N,N-dimethyl-N-(2-phenoxyethyl)-','salt with 4-chlorobenzenesulfonic acid (1:1) (9CI), Ammonium','dimethyl(2-phenoxyethyl)-2-thenyl-','salt with p-chlorobenzenesulfonic acid (1:1) (8CI), Benzenesulfonic acid','4-chloro-','ion(1-)','N,N-dimethyl-N-(2-phenoxyethyl)-2-thiophenemethanaminium (9CI), Benzenesulfonic acid','p-chloro-','ion(1-)','dimethyl(2-phenoxyethyl)-2-thenylammonium (8CI), 611C65, Bancaris, Canopar, Dimethyl(2-phenoxyethyl)-2-thenylammonium closylate, Dimethyl(2-phenoxyethyl)-2-thenylammonium p-chlorobenzenesulfonate, N,N-Dimethyl-N-2-phenoxyethyl-N-2-thenylammonium p-chlorobenzenesulfonate, NSC 106569, Thenium closilate, Thenium closylate, Thenium p-chlorobenzenesulfonate |
| InChI formel | InChI=1S/C15H20NOS.C6H5ClO3S/c1-16(2,13-15-9-6-12-18-15)10-11-17-14-7-4-3-5-8-14;7-5-1-3-6(4-2-5)11(8,9)10/h3-9,12H,10-11,13H2,1-2H3;1-4H,(H,8,9,10)/q+1;/p-1 |
| LEDER | C[N+](C)(CCOc1ccccc1)Cc2cccs2.[O-]S(=O)(=O)c3ccc(Cl)cc3 |
| IUPAC-namn | 4-chlorobenzenesulfonate;dimethyl-(2-phenoxyethyl)-(thiophen-2-ylmethyl)azanium |
| Molekylvikt (g/mol) | 454 |
| Formel vikt | 453.0835 |