Läs mer
Thenyldiamine Hydrochloride, LoGiCal
Produktkod. 30461839
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461839 10 mg
1 options
Produktkod. 30461839 Leverantör LoGiCal Leverantörsnummer LGCFOR1926.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thenyldiamine Hydrochloride
CAS 958-93-0
Analyt- eller komponentnamn Thenyldiamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C14 H19 N3 S . Cl H
Certifikat och efterlevnad ISO 9001
Synonym 1,2-Ethanediamine, N1,N1-dimethyl-N2-2-pyridinyl-N2-(3-thienylmethyl)-, hydrochloride (1:1), 1,2-Ethanediamine, N,N-dimethyl-N'-2-pyridinyl-N'-(3-thienylmethyl)-, monohydrochloride (9CI), N,N-Dimethyl-N'-(3-thenyl)-N'-(2-pyridyl)ethylenediamine hydrochloride, Thenfadil hydrochloride, Thenyldiamine hydrochloride
InChI formel InChI=1S/C14H19N3S.ClH/c1-16(2)8-9-17(11-13-6-10-18-12-13)14-5-3-4-7-15-14;/h3-7,10,12H,8-9,11H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.CN(C)CCN(Cc1ccsc1)c2ccccn2
IUPAC-namn N,N-dimethyl-N'-pyridin-2-yl-N'-(thiophen-3-ylmethyl)ethane-1,2-diamine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 297.85
Kvantitet 10 mg
Formel vikt 297.1066 g/mol