Läs mer
Theophylline-7-acetaldehyde, TRC
Produktkod. 30133168
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133168 50 mg
30136075 250 mg
30008837 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133168 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT34387050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5614-53-9
Molekylformel C9H10N4O3
Molekylvikt (g/mol) 222.2
Synonym 1,2,3,6-Tetrahydro-1,3-dimethyl-2,6-dioxopurine-7-acetaldehyde, 7-(Formylmethyl)theophylline, 7-Theophyllineacetaldehyde
IUPAC-namn 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetaldehyde
LEDER >>O=C1N(C)C(N(C)C2=C1N(CC(O)O)C=N2)=O.O=CCN3C=NC(N(C)C(N4C)=O)=C3C4=O
Specifikationer
Kemiskt namn eller material Theophylline-7-acetaldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C9H10N4O3/c1-11-7-6(8(15)12(2)9(11)16)13(3-4-14)5-10-7/h4-5H,3H2,1-2H3
Formel vikt 222.0753
Procent renhet >95%