missing translation for 'onlineSavingsMsg'
Läs mer

Theophylline-7-acetaldehyde, TRC

Produktkod. 30136075
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136075 250 mg
30133168 50 mg
30008837 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30136075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT343870250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 5614-53-9
Molekylformel C9H10N4O3
Molekylvikt (g/mol) 222.2
Synonym 1,2,3,6-Tetrahydro-1,3-dimethyl-2,6-dioxopurine-7-acetaldehyde, 7-(Formylmethyl)theophylline, 7-Theophyllineacetaldehyde
IUPAC-namn 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetaldehyde
LEDER >>O=C1N(C)C(N(C)C2=C1N(CC(O)O)C=N2)=O.O=CCN3C=NC(N(C)C(N4C)=O)=C3C4=O

Specifikationer

Kemiskt namn eller material Theophylline-7-acetaldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C9H10N4O3/c1-11-7-6(8(15)12(2)9(11)16)13(3-4-14)5-10-7/h4-5H,3H2,1-2H3
Formel vikt 222.0753
Procent renhet >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.