Läs mer
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials and Surfaces
Beskrivning
- Ultra-resistant adhesive ensures no flagging or detachment when stored in liquid nitrogen
- Thermal-transfer printable for the best printout and barcode quality
- Ideal for labeling ambient temperature as well as frozen surfaces without thawing first
- No overlap or wrap-around required for proper adhesion
- Wide temperature range resistance
Specifikationer
Specifikationer
| Färg | White |
| Avsluta | Medium Gloss |
| Formatera | Roll |
| Material | Thermoplastic Film |
| Tryckteknik | Thermal Transfer |
| Produkttyp | Label |
| Sterilitet | Non-sterile |
| Etiketter per rad | 1 etikett |
| Kvantitet | 2000/Pk. |
| Form | Rectangle |
| Visa mer |