Learn More
Brady™ Thermal Transfer Printable Labels, B-427
Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl
Brand:  Brady™ 062361
Additional Details : Weight : 0.40820kg
 View more versions of this product
Product Code. 11552935
  • 2765.00 SEK / 1 roll
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • Available in various sizes
Specifications
Specifications
UL Recognized
Acrylic
Matte
Vinyl, Self-laminating
General and industrial identification
1
Rectangle
-70°C to +70°C
0.102 mm
55.88 x 12.70 mm
55.88 mm
0.5 in.
19.05 mm
Thermal Transfer Printable Labels
White, Translucent
Roll
Thermal Transfer Printable Label
Brady 1244, Brady 1344, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal Transfer Printers, IP300, IP600, BBP81
3000/Pk.
55.88 x 12.7 mm (W x H)
0.004 in.
15.88 mm
2.2 in.
12.70 mm
12.7 mm