Learn More
Brady™ Thermal Transfer Printable Labels, B-427
Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl
Brand: Brady™ 030540
Additional Details : Weight : 1.50590kg
Description
- Available in various sizes
Specifications
UL Recognized | |
Acrylic | |
Matte | |
Vinyl, Self-laminating | |
Cable and wire marking, Panel labeling | |
1 | |
Rectangle | |
-70°C to +70°C | |
0.102 mm | |
80.95 x 48.26 mm | |
48.26 mm | |
80.95 mm |
Thermal Transfer Printable Labels | |
White, Translucent | |
Roll | |
Thermal Transfer Printable Label | |
Brady 1244, Brady 1344, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal Transfer Printers, IP300, IP600, BBP81 | |
1500/Pk. | |
48.26 x 80.95 mm (W x H) | |
0.004 in. | |
54.61 mm | |
1.9 in. | |
3.187 in. | |
19.05 mm |