Learn More
Corning™ ThermalTray™ Thermo-conductive Platforms
Support Corning CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths, and liquid nitrogen.
3650.00 SEK - 4070.00 SEK
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
15582841
|
Corning™
432073 |
9L ice pan with liquid nitrogen | SLP, Slim, Low Profile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15592841
|
Corning™
432074 |
9L ice pan with crushed ice | LP, Low Profile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15502851
|
Corning™
432075 |
Water bath | HP, High Profile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Corning ThermalTray™ thermo-conductive platforms support CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.Specifications
ThermalTray Thermo-conductive platform |