Learn More
Corning™ ThermalTray™ Thermo-conductive Platforms
Support Corning CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths, and liquid nitrogen.
3650.00 SEK - 4070.00 SEK
Specifications
Description ThermalTray Thermo-conductive platform
View More Specs
Products 3
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
15582841
View Documents Promotion Details
Corning™
432073
9L ice pan with liquid nitrogen SLP, Slim, Low Profile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15592841
View Documents Promotion Details
Corning™
432074
9L ice pan with crushed ice LP, Low Profile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15502851
View Documents Promotion Details
Corning™
432075
Water bath HP, High Profile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Corning ThermalTray™ thermo-conductive platforms support CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Specifications
    Specifications
    ThermalTray Thermo-conductive platform