Läs mer
Thermo Scientific™ 24-Well Semi-Skirted PCR Plate
Produktkod. p-4505416
Change view
Färg:
Gult
Rött
Vitt
Förpackningsstorlek:
Paket med 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Färg unitSize
10563795 Rött Paket med 50
10405353 Gult Paket med 50
12657506 Vitt Paket med 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 10563795 Leverantör Thermo Scientific™ Leverantörsnummer AB0624/R
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
24-well semi-skirted plate for PCR and qPCR applications.
The 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
The Thermo Scientific 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
Highlights
• Conveniently pre-cut into 24-well segments
• Suitable for use in 0.2 mL thermal cycler blocks
• Semi-skirt adds rigidity and allows for labeling or barcoding
• 0.3 mL maximum well volume (when used with adhesive seals)
• Sealable using adhesive films and foils and all cap strips
Certifikat
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifikationer
Formatera Standard
Array 4 x 6
Färg Rött
Material Virgin Polypropylene
Antal Wells 24