Learn More
Thermo Scientific™ 384-Well Full-Skirted Plates, Standard
Fully-skirted 384-well standard plates compatible with a wide range of block thermal cyclers for use in PCR and qPCR applications
Brand: Thermo Scientific™ AB-1384/K
Additional Details : Weight : 1.08000kg
Description
Designed for high-throughput PCR, the Thermo Scientific 384-well plate is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40μL and a working volume of 25μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.
Highlights
- Fully skirted for use with automated systems
- Compatible with all leading 384 block thermal cyclers
- 40μL maximum well volume, 25μL working volume
- Raised rim design around each well enables secure sealing and safeguards against evaporation
- Uniform, thin well walls deliver maximum and consistent heat transfer
- White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
- Cut corner: A24
Certifications
Certifed free from human DNA, RNases and DNases.
Specifications
Certifed free from human DNA, RNases and DNases | |
16 x 24 | |
Virgin Polypropylene | |
50/Pk. |
Standard | |
Black | |
384 |