Learn More
Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples
8641.72 SEK - 9411.54 SEK
Specifications
Column Format | Capillary Column |
---|---|
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 100 Å |
Surface Area | 450 m²/g |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15142428
|
Thermo Scientific™
164901 |
50 mm | 0.3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15132428
|
Thermo Scientific™
164900 |
150 mm | 1 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15152428
|
Thermo Scientific™
164902 |
150 mm | 0.3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- High resolution in protein identification, biomarker discovery, and systems biology
- Particular loadability allows hghest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
- Inside diameter 300μm
Specifications
Capillary Column | |
2 to 8 | |
450 m²/g | |
C18 | |
Silica | |
Reversed Phase |
5 μm | |
100 Å | |
L1 | |
nanoLC | |
15% | |
Reversed Phase |