Learn More
Thermo Scientific™ Barnstead™ Deionization System Disposable Filters
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII
Brand: Thermo Scientific™ FL703X2
Additional Details : Weight : 0.02160kg
Description
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII
Specifications
Autoclavable | |
0.2 μm | |
Filling bell |
1 | |
Reverse osmosis and TII water purification system | |
Deionization System Disposable Filter |