Learn More
Thermo Scientific™ Barnstead™ Water Purification System Replacement Cartridges
Product Code. 13204749 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cartridge
Organicfree Cartridge
Pretreatment Cartridge
TFM Membrane
For Use With (Equipment):
Easypure Reverse Osmosis System
Nanopure Infinity System
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
13204749 Pretreatment Cartridge Nanopure Infinity System Each
11810810 Cartridge Nanopure Infinity System Each
11820810 Organicfree Cartridge Nanopure Infinity System Each
11810920 TFM Membrane Easypure Reverse Osmosis System Each
10080614 Pretreatment Cartridge Easypure Reverse Osmosis System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13204749 Supplier Thermo Scientific™ Supplier No. D50251
  • ONLINE EXCLUSIVE PRICE
    2715.00 SEK
    2659.57 SEK / Paket med 1
    Save 55.43 SEK
    2% Av
 Restricted Item
Beräknad leverans: 18-02-2027
For Easypure™ RO water purification system
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Deionized, Tap
Resin Type Activated Carbon and Macroreticular
For Use With (Equipment) Nanopure Infinity System
Type Pretreatment Cartridge