Learn More
Unity Lab Services™ Thermo Scientific™ Liquid Nitrogen Storage Equipment Validation Service
Product Code. 15681599
Product Code. 15681599 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003811
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Beräknad leverans: 25-01-2027
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Liquid Nitrogen Storage Equipment Validation Service for Thermo Scientific CryoExtra™ High-Efficiency Cryogenic Storage Systems Models: CE8180, CE8120, CE8140, CE8194
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
  • Key Tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Liquid Nitrogen Storage Equipment
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing.
Product Type IQ/OQ Service
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service