Learn More
Thermo Scientific™ MIC 2000 Microtiter microplate
For support of microbiolgical applications
Brand: Thermo Scientific™ 12-0151
Additional Details : Weight : 4.33000kg
Description
These products are suitable for applications including bacterial growth, bacterial identification, minimum inhibitory concentration (MIC), hybridization, hybridoma plating and cloning.
- Microtiter microplate format
- Polystyrene
- U-bottom
Order Info
Bag of 20; Box of 100
Specifications
Without Lid | |
Polystyrene | |
Sterile | |
0.28 mL |
Clear | |
96 | |
Non-treated | |
Round Bottom (U-well) |