Learn More
Thermo Scientific™ Nunc™ Caps for TufRol™ Roller Bottles
Vented and non-vented
Brand:  Thermo Scientific™ 112250
Additional Details : Weight : 6.50000kg
 View more versions of this product
Product Code. 11997964
  • 3481.00 SEK / Pack of 500
Estimated Shipment: 08-12-2021
Add to basket
Description
Description
Vented and non-vented
  • Molded of high density polyethylene (HDPE)
  • Designed to prevent misthreading and to provide a secure seal
  • Vented cap has an integral hydrophobic 0.2μm microporous membrane filter vent, allowing consistent gas exchange when cap is in the closed position
Specifications
Specifications
Non-vented caps
TufRol roller bottle
500 Cs.