Learn More
Invitrogen™ Mouse IL-7 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8664
Additional Details : Weight : 0.01000kg
Description
IL-7 (mouse) Recombinant Protein, RP-8664, from Invitrogen.
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells. Recombinant mouse soluble RANK Ligand is a non-glycosylated protein, containing 174 amino acids, with a molecular weight of 19.9 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to induce osteoclast formation on mouse RAW264.7 cells using a concentration of 5.0-10.0ng/mL. The specific activity of this protein is 2x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: PAMMEGSWLD VAQRGKPEAQ PFAHLTINAA SIPSGSHKVT LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPT DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISIQV SNPSLLDPDQ DATYFGAFKV QDID.Specifications
P10168 | |
Control | |
16196 | |
Mouse IL-7 | |
E. coli | |
RUO | |
Mouse IL-7 | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
0.1mg/mL | |
0.1%TFA with no preservative | |
∽15 | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
M-IL-7 | |
Il7 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only