missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Ubiquitin, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ LFP0041
Additional Details : Weight : 0.99000kg
Description
Ubiquitin Recombinant Protein, LF-P0041, from Invitrogen.
Ubiquitin (Ub) is a small protein that is composed of 76 amino acids. Ub is found only in eukaryotic organisms and is highly conserved. Ub can exist either in free form or as part of a complex with other proteins. In the latter case, Ub is attached (conjugated) to proteins through a covalent bond between the glycine at the C-terminal end of Ub and the side chains of lysine on the proteins; Ubiquitination. Ub functions to regulate protein turnover in a cell by closely regulating the degradation of specific proteins. Ubiquitin has been immunohistochemically localized to a number of pathological inclusions, including Lewy bodies of Parkinson′s disease, neurofibrillary of Alzheimer′s disease, Pick bodies of Pick′s disease. This protein corresponds to human Ubiquitin (1-76 aa with N-GST tag), which has a molecular weight of 35 kD. Purity: >95% by SDS-PAGE. After reconstitution, store at -80°C and avoid freeze and thaw cycles.Specifications
P62979 | |
0.02M HEPES with no preservative; pH 7.4 | |
Human Ubiquitin | |
500μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human Ubiquitin | |
Human | |
Hu | |
Recombinant | |
Recombinant |
Control | |
6233 | |
7.4 | |
E. coli | |
RUO | |
RPS27A | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only.