missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse VEGF-165 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8672
Additional Details : Weight : 0.01000kg
Description
Vascular Endothelial Growth Factor 165 (mouse) Recombinant Protein, RP-8672, from Invitrogen.
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several variants, VEGF-165 being the most abundant. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%. Recombinant mouse Exodus-2 is a non-glycosylated protein, containing 110 amino acids, with a molecular weight of 12 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract 5-10 day old cultures of human peripheral blood lymphocytes at concentrations between 200 ng/mL- 600 ng/mL. The specific activity of this protein is 5x10^3 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FSPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG.Specifications
Q00731 | |
Control | |
22339 | |
Mouse VEGF-165 | |
E. coli | |
RUO | |
Mouse VEGF-165 | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
0.1mg/mL | |
0.1%TFA with no preservative | |
∽39 | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
M-VEGF-165 | |
Vegfa | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only.