missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant SC250EXP SpeedVac Kits

Large-capacity SpeedVac kits—everything you need to effectively concentrate high boiling point solvents

Brand:  Thermo Scientific™ SC250EXPA-230


Additional Details : Weight : 127.00000kg

Product Code. 11831214

  • 429740.00 SEK / Each

Estimated Shipment: 16-09-2022
to see stock.
Add to basket

Description

Description

Dual vacuum control on our SPD250EXP provides flexibility to use many sample types.

  • Four IR heat lamps for drying high boiling point solvents such as DMSO and DMF
  • Dual vacuum control of RAMP and LEVEL eliminates samples from bumping and/or freezing, providing optimization for various sample types
  • Kalrez™ and PTFE-coated materials along the entire vapor path for maximum chemical resistance and durabilityi
  • Dual valve mechanism for improved durability and exhausting
  • Comes complete with enlarged vacuum port, extra wide tubing and new extra large mouth glass condensation flask
  • Heated glass lid reduces drying times and prevents condensation
  • Digital display of parameters (time, temperture and vacuum)
  • Standard glass safety lids

Solvent and Solvent Combinations - SC250P1

  • Acetic Acid
  • Acetone
  • Ammonium Hydroxide
  • Formic Acid
  • Hydrochloric Acid
  • TFA
  • Methylane Chloride and Water
  • Drug Metabolites in SPE/Urine
  • DCM and Hepatane
  • Methylene Chloride and MEOH
  • THF/Pyridine (95:5)
  • DMSO and Methanol
Solvent and Solvent Combinations - SC250P2
  • Chloroform
  • DMSO
  • DMF
  • Ethyl Acetate
  • Hexane
  • Toluene
  • Methanol and Chloroform

SC250P1

  • Solid phase extractions
  • Drying lipid extracts
  • Flash chromatography fractions
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
SC250P2
  • Complete management of evaporating dissolution solvents

Specifications

Specifications

60 Hz
115 V
115 V 60 Hz
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers