Learn More
Epredia™ Shandon™ TissueLoc™ Biopsy Cassettes IV in Tube Packs
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
Brand: Epredia™ 41750-T
Additional Details : Weight : 2.50000kg
Description
- Available in an assortment of colors
- Packaged in 10 tubes of 75 cassettes each
Specifications
Hinged | |
White | |
Biopsy Cassette |