Läs mer
Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Produktkod. p-147654
Kapacitet (metrisk):
2 l
Spänning:
208 V
Controllertyp:
Åttastegs-/ettprogramsstyrning
Produktkod. 10447643
Brand: Thermo Scientific™ F4792080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
Power cord (-60 models include 20-amp cord; -33 units available with plug for China)
Compact, lightweight benchtop furnaces
  • For added protection, the door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to heating elements, preventing a thermocouple failure runaway condition
  • Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
  • Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
  • 0.95cm (0.38 in.) dia. port for monitoring chamber temperatures with independent measuring device at rear of chamber
F47900 Models
  • 2L (0.07 cu. ft.) of heating area
  • F48000 Models
    • 5.9L (0.20 cu. ft.) of heating area
    • Includes ceramic shelf (SH480X1) that doubles furnace load capacity
    A1 Controller
    • Digital single setpoint control
    • Single display shows actual temperature or setpoint
    B1 Controller
    • Digital single setpoint control with a single ramp to setpoint and a dwell
    • Single display shows actual temperature or setpoint
    • Furnaces with this control also use a mechanical Overtemperature Protection relay
    C1 Controller
    • Digital programmable control with one stored program of eight segments
    • Furnaces with this control also use a mechanical Overtemperature Protection relay
    D1 Controller
    • Digital programmable control with four stored programs, 16 segments per program
    • With RS-232 communications interface
    • Furnaces with this control also use a mechanical Overtemperature Protection relay
    Certifikat
    CSA approved, CE marked as indicated
    TRUSTED_SUSTAINABILITY
    Specifikationer
    Controllertyp Åttastegs-/ettprogramsstyrning
    Kapacitet (metrisk) 2 l
    Temperatur (metrisk) 100°C to 1200°C
    Spänning 208 V
    Wattal 1560 w
    Amperage 7.5 A
    Frekvens 50/60 Hz