Läs mer
Thermo Scientific™ Thermolyne™ Industrial Benchtop Muffle Furnaces
Produktkod. 15961437
Energiförbrukning:
1560 v
2230 W
Temperaturkontroll:
Digitalt med 1 program/8 segment
Spänning:
240 V
Förpackningsstorlek:
Varje
Produktkod. 15961437
Brand: Thermo Scientific™ FD1540M33
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Perform ultra-high temperature applications with this industrial benchtop muffle furnace, which reaches a maximum temperature of 1200°C.
Perform ultra-high temperature applications with the Thermo Scientific™ Thermolyne™ Industrial Benchtop Muffle Furnace, which reaches a maximum temperature of 1200°C. Unit includes multiple safety features and choice of two temperature-control options. Heavy-duty firebrick insulation surrounds the opening for added durability.
  • Adjustable Alarm or Overtemperature Protection (OTP) setting can be used to protect the furnace or loaded chamber from excessive heat
  • Thermocouple break protection cuts power to the heating elements, preventing a thermocouple failure runaway condition
  • Counter-weighted door swings upward, directing heat away from operator
  • Heavy-duty firebrick insulation surrounds chamber opening for added durability
  • Four individual embedded elements in special refractory cement permit excellent heat distribution in the chamber
  • Door safety switch protects operator by removing power to the heating elements upon opening the door
  • Rear-mounted 0.38in. (0.95cm) diameter port for monitoring chamber temperatures with independent measuring devices
  • LED display simultaneously shows both setpoint and actual furnace temperatures in either °C or °F (programmable models only)
Single-setpoint Controller (B1)
  • Single ramp to setpoint and a dwell
  • Single display shows actual temperature or setpoint
8-step Programmable Controller (C1)
  • Ramps to vary heat-up rate and dwell cycles to hold temperature at set levels from 0.1 to 999.9 minutes
  • “Holdback” feature “holds” program until furnace temperature heats up or cools down to preprogrammed parameters—program will never outrun furnace performance
Includes:
Oven, Platinel™ II thermocouple and a ceramic hearth plate to protect the bottom heating element.
Note:
120V models require fixed wiring and are not supplied with a power cord.
TRUSTED_SUSTAINABILITY
Specifikationer
Kapacitet (engelska) 0.08 cu.ft.
Certifikat och efterlevnad All units cUL, UL listed; -33 units also CE marked.
Kapacitet (metrisk) 2.2 L
Temperatur (metrisk) 100°C to 1200°C
Typ Multisegment, Single Program
Energiförbrukning 1560 v
Temperaturkontroll Digitalt med 1 program/8 segment
Mått (D x B x H) Inredning 9 x 4 x 3.75 in. (22.8 x 10.1 x 9.5 cm)
Mått (L x B x H) Exteriör 18 x 11 x 16.5 in. (45.7 x 27.9 x 41.9 cm)
Längd (engelska) 18 in.
Längd (metrisk) 45.7 cm
Bredd (engelska) 4 in.
Bredd (metrisk) 27.9
Bredd (metrisk) Interiör 10.1 cm
Höjd (engelska) 16.5 in.
Höjd (engelska) Interiör 3.75 in.
Höjd (metrisk) 41.9 cm
Höjd (metrisk) interiör 9.5 cm
Depth (engelska) Interiör 9 in.
Djup (metrisk) interiör 22.8 cm
Fraktvikt (engelska) 52 lb.
Fraktvikt (metrisk) 23.5 kg
Elektriska krav 240 V 50/60 Hz
Spänning 240 V
Amperage 6.5 A
Frekvens 50/60 Hz
Visa mer Visa mindre
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.