Beskrivning
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
- Shielded thermoblocks are contact-safe
- Efficient exchange simplifies operation
- Tubes are visible during operation and easily accessible
Specifikationer
Specifikationer
| För användning med (utrustning) | Eppendorf-röret |
| Håller | 8 x 5,0 ml |
| Typ | Block |
| För användning med (applikation) | For 8 x Eppendorf Tube 5.0 mL |
| Inkluderar | Not Applicable |