Learn More
Quickfit™ Thermoplastic Polyester Tubing Clamps
For use with flexible rubber and plastic tubing
Brand: Quickfit KT14
452.20 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.08000kg
Description
For use with flexible rubber and plastic tubing
- Provides fine flow control of liquids and gases
- Easy one handed adjustment: just roll serated wheel along sloped floor
- Manufactured from thermoplastic polyester
Specifications
Tubing Clamp | |
Thermoplastic Polyester | |
Up to 14mm O.D. tubing | |
Used with flexible Rubber and Plastic tubing |