Läs mer
theta-Cypermethrin, TRC
Produktkod. 30008536
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008536 100 mg
30003323 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008536 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC989525100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71697-59-1
Molekylformel C22 H19 Cl2 N O3
Molekylvikt (g/mol) 416.3
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1α(S*),3β]-(±)-, NRDC 159, Theta-cypermethrin, Transmix
IUPAC-namn [(R)-cyano-(3-phenoxyphenyl)methyl] (1S,3R)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
LEDER CC1(C)[C@@H](C=C(Cl)Cl)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2
Specifikationer
Kemiskt namn eller material theta-Cypermethrin
Kvantitet 100 mg
InChI formel InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18-,20+/m0/s1
Formel vikt 415.0742