Läs mer
Thiamine Disulfide Hydrate, TRC
Produktkod. 30454665
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30454665 5 g
1 options
Produktkod. 30454665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3441405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiamine Disulfide Hydrate
CAS 1899166-52-9
Molekylformel C24H34N8O4S2.H2O
Kvantitet 5 g
Synonym N,N'-[Dithiobis[2-(2-hydroxyethyl)-1-methyl-2,1-ethenediyl]]bis[N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]formamide, Aktivin, Algoneurina, Alitia S, Aneurin Disulfide, Aneurine Disulfide, Apren S, Daiomin, Daisazin, Feidmin 5, Neolamin, SSB1, TDS, Thiamidin F, Thiamin Disulfide, Vitamin B1 Disulfide
InChI formel InChI=1S/C24H34N8O4S2.H2O/c1-15(31(13-35)11-19-9-27-17(3)29-23(19)25)21(5-7-33)37-38-22(6-8-34)16(2)32(14-36)12-20-10-28-18(4)30-24(20)26;/h9-10,13-14,33-34H,5-8,11-12H2,1-4H3,(H2,25,27,29)(H2,26,28,30);1H2
LEDER O.CC(=C(CCO)SSC(=C(C)N(Cc1cnc(C)nc1N)C=O)CCO)N(Cc2cnc(C)nc2N)C=O
IUPAC-namn N-[(4-amino-2-methyl-pyrimidin-5-yl)methyl]-N-[2-[[2-[(4-amino-2-methyl-pyrimidin-5-yl)methyl-formyl-amino]-1-(2-hydroxyethyl)prop-1-enyl]disulfanyl]-4-hydroxy-1-methyl-but-1-enyl]formamide;hydrate
Molekylvikt (g/mol) 580.72
Formel vikt 580.225