Läs mer
Thiamine Sulfate, TRC
Produktkod. 30009729
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009729 1 g
30012660 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3440851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2380-61-2
Molekylformel C12H16N4O4S2
Molekylvikt (g/mol) 344.41
Synonym 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(sulfooxy)ethyl]thiazolium Inner Salt, Thiamine Hydrogen Sulfate Ester Inner Salt;
IUPAC-namn 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl sulfate
LEDER NC1=NC(C)=NC=C1C[N+]2=CSC(CCOS(=O)([O-])=O)=C2C
Specifikationer
Kemiskt namn eller material Thiamine Sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C12H16N4O4S2/c1-8-11(3-4-20-22(17,18)19)21-7-16(8)6-10-5-14-9(2)15-12(10)13/h5,7H,3-4,6H2,1-2H3,(H2-,13,14,15,17,18,19)
Formel vikt 344.0613
Procent renhet >95%