missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 61°C to 65°C |
Color | Brown |
Quantity | 250 mg |
Beilstein | 107137 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 113.13 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | Thiazole-4-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'