missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-7055447
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
3364-80-5
C4H3NOS
113.13
MFCD00626896
WRFKSVINLIQRKF-UHFFFAOYSA-N
thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole
2763214
1,3-thiazole-4-carbaldehyde
O=CC1=CSC=N1
This item is not returnable. View return policy
3364-80-5
C4H3NOS
113.13
MFCD00626896
WRFKSVINLIQRKF-UHFFFAOYSA-N
thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole
2763214
1,3-thiazole-4-carbaldehyde
O=CC1=CSC=N1

Thiazole-4-carboxaldehyde, 95%, C4H3NOS, CAS Number-3364-80-5, 4-formylthiazole, 4-thiazolecarbaldehyde, 1,3-thiazole-4-carboxaldehyde, 4-formyl-1,3-thiazole, thiazole-4-carbaldehyde, thiazole-4-carboxyaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxaldehyde, pubchem13812, zlchem 1299, 5g

Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 61°C to 65°C
Color Brown
Quantity 250 mg
Beilstein 107137
Solubility Information Slightly soluble in water.
Formula Weight 113.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Thiazole-4-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'