Läs mer
Thiazolylacetylglycine Oxime, TRC
Produktkod. 30451285
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30451285 50 mg
1 options
Produktkod. 30451285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT34423050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiazolylacetylglycine Oxime
CAS 178949-03-6
Molekylformel C7 H8 N4 O4 S
Kvantitet 50 mg
Synonym (Z)-N-[(2-Amino-4-thiazolyl)(hydroxyimino)acetyl]glycine, N-[(2Z)-(2-Amino-4-thiazolyl)(hydroxyimino)acetyl]glycine, Cefdinir Thiazolylacetyl Glycine Oxime Impurity (USP)
InChI formel InChI=1S/C7H8N4O4S/c8-7-10-3(2-16-7)5(11-15)6(14)9-1-4(12)13/h2,15H,1H2,(H2,8,10)(H,9,14)(H,12,13)/b11-5-
LEDER Nc1nc(cs1)\C(=N\O)\C(=O)NCC(=O)O
IUPAC-namn 2-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]acetic acid
Molekylvikt (g/mol) 244.23
Formel vikt 244.0266