missing translation for 'onlineSavingsMsg'
Läs mer

STEMCELL Technologies Thiazovivin

Produktkod. 17190042
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17190042 1 mg
17100052 5 mg
17129971 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17190042 Leverantör STEMCELL Technologies Leverantörsnummer 72252

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

RHO/ROCK pathway inhibitor; Inhibits ROCK

  • Thiazovivin is a selective inhibitor of Rho-associated coiled-coil containing protein kinase (ROCK), a serine/threonine kinase that plays a role in cell polarity, contraction, and actin cytoskeleton reorganization (Xu et al.) Thiazovivin is effective at 5-fold-lower concentrations than another common ROCK inhibitor Y-27632 (Catalog #72302; Xu et al.)
  • Promotes survival of human embryonic stem (ES) cells during dissociation by stabilizing E-cadherin and improves cell attachment (Xu et al.)
  • Promotes survival of single human induced pluripotent stem (iPS) cells during transfection for TALEN-mediated genome editing (Sun and Zhao)
  • Increases the efficiency of reprogramming human somatic cells to iPS cells, in combination with PD0325091 and SB431542 (Lin et al.)
  • Increases the efficiency of reprogramming human cord blood mononuclear cells to iPS cells (Hu et al.)

Specifikationer

CAS 1226056-71-8
Synonym Tzv
Kvantitet 1 mg
Molekylformel C15H13N5OS
Renhet ≥98%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.