Läs mer
Thienamycin (>80%), TRC
Produktkod. 30456173
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456173 10 mg
30450173 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456173 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT34427010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59995-64-1
Molekylformel C11 H16 N2 O4 S
Molekylvikt (g/mol) 272.32
Synonym 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[(2-aminoethyl)thio]-6-[(1R)-1-hydroxyethyl]-7-oxo-, (5R,6S)-, 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[(2-aminoethyl)thio]-6-(1-hydroxyethyl)-7-oxo-, [5R-[5α,6α(R*)]]-, (5R,6S)-3-[(2-Aminoethyl)thio]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, (+)-Thienamycin, Thienamycin
IUPAC-namn (5R,6S)-3-(2-aminoethylsulfanyl)-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
LEDER C[C@@H](O)[C@@H]1[C@H]2CC(=C(N2C1=O)C(=O)O)SCCN
Specifikationer
Kemiskt namn eller material Thienamycin
Kvantitet 10 mg
InChI formel InChI=1S/C11H16N2O4S/c1-5(14)8-6-4-7(18-3-2-12)9(11(16)17)13(6)10(8)15/h5-6,8,14H,2-4,12H2,1H3,(H,16,17)/t5-,6-,8-/m1/s1
Formel vikt 272.0831