Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thifensulfuron-methyl |
| CAS | 79277-27-3 |
| CAS Max % | ≤ 100% |
| Densitet | 1.49 g/mL |
| Beilstein | 7448062 |
| Färg | White |
| Smältpunkt | 180°C |
| Synonym | Methyl 3-(4-methoxy-6-methyl-1,3,5-triazin-2-ylcarbamoylsulfamoyl)thiophene-2-carboxylate |
| Molekylvikt (g/mol) | 387.39 g/mol |
| Kvantitet | 100 mg |
| Visa mer |