Läs mer
Thifensulfuron-methyl, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thifensulfuron-methyl |
| CAS | 79277-27-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H13 N5 O6 S2 |
| Kvantitet | 5 g |
| Synonym | DPX-M 6316, Harmony, Harmony 75DF, Harmony GT, Harmony GT-XP, Pinnacle, Pinnacle 25DF, Refine, Refine DF, Thiameturon-methyl, Thifensulfuron methyl, Thifensulfuron methyl ester, 2-Thiophenecarboxylic acid','3-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-','methyl ester, Thifensulfuron Methyl |
| InChI formel | InChI=1S/C12H13N5O6S2/c1-6-13-10(16-12(14-6)23-3)15-11(19)17-25(20,21)7-4-5-24-8(7)9(18)22-2/h4-5H,1-3H3,(H2,13,14,15,16,17,19) |
| LEDER | COC(=O)c1sccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2 |
| IUPAC-namn | methyl 3-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]thiophene-2-carboxylate |
| Molekylvikt (g/mol) | 387.39 |
| Visa mer |