Läs mer
Thioflavine S (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thioflavine S (Technical Grade) |
| CAS | 1326-12-1 |
| Molekylformel | C17H19N2S.Cl |
| Kvantitet | 5 g |
| Synonym | Amanil Brilliant Yellow S, Atlantic Brilliant Flavine S, C.I. 49010, Chloramine Brilliant Flavine S, Chromine G, Diazol Brilliant Yellow S, Direct Brilliant Flavine S, Direct Brilliant Yellow 3GN-CF, Direct Brilliant Yellow 5GX, Direct Flavine S, Direct Pure Yellow TS, Eljon Ex Yellow Green Toner, Erie Flavine S, Methylated Sulfonated Primuline Base, Phenamine Brilliant Yellow 3G, Thioflavin S |
| InChI formel | InChI=1S/C17H19N2S.ClH/c1-12-5-10-15-16(11-12)20-17(19(15)4)13-6-8-14(9-7-13)18(2)3;/h5-11H,1-4H3;1H/q+1;/p-1 |
| LEDER | N[C@H](CC1=CC=C(CC)C=C1)C(O)=O |
| Molekylvikt (g/mol) | 318.9 |