Läs mer
ThioGlo1, TRC
Produktkod. 30454443
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454443 50 mg
30452745 100 mg
30451096 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454443 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT35076550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137350-66-4
Molekylformel C20H13NO7
Molekylvikt (g/mol) 379.32
Synonym Methyl 10-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-9-methoxy-3-oxo-3H-benzo[f]chromene-2-carboxylate
IUPAC-namn methyl 10-(2,5-dioxopyrrol-1-yl)-9-methoxy-3-oxobenzo[f]chromene-2-carboxylate
LEDER C1(=CC=C2C(=C1N3C(C=CC3=O)=O)C4=C(C=C2)OC(C(=C4)C(=O)OC)=O)OC
Specifikationer
Kemiskt namn eller material ThioGlo1
Kvantitet 50 mg
InChI formel InChI=1S/C20H13NO7/c1-26-14-6-4-10-3-5-13-11(9-12(19(24)27-2)20(25)28-13)17(10)18(14)21-15(22)7-8-16(21)23/h3-9H,1-2H3
Formel vikt 379.0692