Läs mer
Thioglycolic Acid 2-Ethylhexyl Ester, TRC
Produktkod. 30006510
Change view
Kvantitet:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006510 10 mL
30008903 1 mL
30004033 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006510 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT35076310ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7659-86-1
Molekylformel C10H20O2S
Molekylvikt (g/mol) 204.33
Synonym 2-Ethylhexyl 2-Mercaptoacetate, 2-Ethylhexyl 2-Sulfanylacetate, 2-Ethylhexyl Mercaptoacetate, 2-Ethylhexyl Thioglycolate, 2-Mercaptoacetic Acid 2-Ethylhexyl Ester, EHTG, Mercaptoacetic Acid 2-Ethylhexyl Ester;
IUPAC-namn 2-ethylhexyl 2-sulfanylacetate
LEDER CCCCC(CC)COC(=O)CS
Specifikationer
Kemiskt namn eller material Thioglycolic Acid 2-Ethylhexyl Ester
Kvantitet 10 mL
InChI formel InChI=1S/C10H20O2S/c1-3-5-6-9(4-2)7-12-10(11)8-13/h9,13H,3-8H2,1-2H3
Formel vikt 204.1184