Läs mer
Thiometon Sulfoxide, TRC
Produktkod. 30449420
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30449420 250 mg
1 options
Produktkod. 30449420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT344675250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiometon-sulfoxide
CAS 2703-37-9
Anteckningar om renhetsgrad HPLC
Molekylformel C6 H15 O3 P S3
Kvantitet 250 mg
Synonym Intrathion sulfoxide, Intration sulfoxide, O,O-Dimethyl S-(2-ethylsulfinyl)ethyl dithiophosphate, O,O-Dimethyl S-(2-ethylsulfinylethyl) phosphorodithioate, Oxythiometon, Thiometon sulfoxide, Thiometon-sulfoxide
InChI formel InChI=1S/C6H15O3PS3/c1-4-13(7)6-5-12-10(11,8-2)9-3/h4-6H2,1-3H3
LEDER CCS(=O)CCSP(=S)(OC)OC
IUPAC-namn 2-ethylsulfinylethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane
Molekylvikt (g/mol) 262.35
Formel vikt 261.9921