Läs mer
Thionaphthenquinone, TRC
Produktkod. 30454854
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454854 10 g
30450279 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT36769010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 493-57-2
Molekylformel C8 H4 O2 S
Molekylvikt (g/mol) 164.18
IUPAC-namn 1-benzothiophene-2,3-dione
LEDER O=C1Sc2ccccc2C1=O
Specifikationer
Kemiskt namn eller material Thionaphthenquinone
Kvantitet 10 g
InChI formel InChI=1S/C8H4O2S/c9-7-5-3-1-2-4-6(5)11-8(7)10/h1-4H
Formel vikt 163.9932