Läs mer
Thioquinox, TRC
Produktkod. 30455564
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30455564 250 mg
1 options
Produktkod. 30455564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT368298250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thioquinox
CAS 93-75-4
Molekylformel C9 H4 N2 S3
Kvantitet 250 mg
Synonym 1,3-Dithiolo[4,5-b]quinoxaline-2-thione (9CI','ACI), Carbonic acid','trithio-','cyclic 2,3-quinoxalinediyl ester (8CI), Carbonic acid','trithio-','cyclic ester with 2,3-quinoxalinedithiol (6CI), 2,3-Quinoxalinedithiol cyclic trithiocarbonate, 2-Thioxo-1,3-dithiolo[4,5-b]quinoxaline, 2H-[1,3]Dithiolo[4,5-b]quinoxaline-2-thione, Bayer 30686, Chinothionat, Eradex, Quinothionate, Quinoxaline-2,3-diyl trithiocarbonate, Readex, Thiaquinox, Thioquinox
InChI formel InChI=1S/C9H4N2S3/c12-9-13-7-8(14-9)11-6-4-2-1-3-5(6)10-7/h1-4H
LEDER S=C1Sc2nc3ccccc3nc2S1
IUPAC-namn [1,3]dithiolo[4,5-b]quinoxaline-2-thione
Molekylvikt (g/mol) 236.34
Formel vikt 235.9537