Läs mer
Thioridazine Disulfoxide, TRC
Produktkod. 30450870
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30450870 10 mg
1 options
Produktkod. 30450870 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT53926010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mesoridazine 5-Sulfoxide
CAS 53926-89-9
Molekylformel C21 H26 N2 O2 S2
Kvantitet 10 mg
Synonym 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine 5-Oxide, Phenothiazine','10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)-','5-oxide (7CI), Thioridazine 2,5-disulfoxide, Thioridazine disulfoxide, Mesoridazine 5-Sulfoxide
InChI formel InChI=1S/C21H26N2O2S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)27(25)21-11-10-17(26(2)24)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3
LEDER CN1CCCCC1CCN2c3ccccc3S(=O)c4ccc(cc24)S(=O)C
IUPAC-namn 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine 5-oxide
Molekylvikt (g/mol) 402.57
Formel vikt 402.1436